missing translation for 'onlineSavingsMsg'
Learn More

CHRNA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26025-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16844555

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CHRNA9 is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor superfamily. CHRNA9 is a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. It is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea.This gene is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor gene superfamily. It encodes a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. This protein is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea. The protein is additionally expressed in keratinocytes, the pituitary gland, B-cells and T-cells.
Specifications

Specifications

CHRNA9
Polyclonal
Unconjugated
CHRNA9
CHRNA9, HSA243342, NACHR alpha 9, NACHRA9
Rabbit
Antigen Affinity Chromatography
RUO
231252, 55584
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UGM1
CHRNA9
CHRNA9 Fusion Protein Ag23272
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers