Learn More
Proteintech CHORDC1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25525-1-AP-150UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17221376
  • 3828.53 NOK / 150µL
Estimated Shipment: 20-06-2023
Description
The CHORDC1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CHORDC1. This antibody recognizes human antigen. The CHORDC1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
CHORDC1
Polyclonal
Unconjugated
150 μL
26973
Liquid
Western Blot, ELISA
600 μg/mL