missing translation for 'onlineSavingsMsg'
Learn More

CHORDC1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. p-7231501 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17231376 20 μL 20µL
17221376 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17231376

Brand: Proteintech 255251AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CHORDC1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CHORDC1. This antibody recognizes human antigen. The CHORDC1 antibody has been validated for the following applications: WB,ELISA analysis.

Specifications

Antigen CHORDC1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 600 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 26973
Target Species Human
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.