Learn More
Chloroacetaldehyde diethyl acetal, 98%
Product Code. 15478869
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478869 500 g 500g
15468869 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15478869 Supplier Thermo Scientific Alfa Aesar Supplier No. L07661.36
 Restricted Item
In Stock
Add to basket
CAS: 621-62-5 | C6H13ClO2 | 152.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 621-62-5
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000949
InChI Key OVXJWSYBABKZMD-UHFFFAOYSA-N
Synonym chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
PubChem CID 12128
IUPAC Name 2-chloro-1,1-diethoxyethane
SMILES CCOC(CCl)OCC
Specifications
Melting Point -45°C
Density 1.018
Boiling Point 156°C to 157°C
Flash Point 50°C (122°F)
Linear Formula ClCH2CH(OCH2CH3)2
Refractive Index 1.416
Quantity 500 g
UN Number UN1993
Beilstein 969254
Sensitivity Moisture sensitive
Formula Weight 152.62
Percent Purity 98%
Chemical Name or Material Chloroacetaldehyde diethyl acetal