accessibility menu, dialog, popup

UserName

CAS RN 621-62-5

H3COClOCH3

CAS RN 621-62-5

IUPAC Name: 2-chloro-1,1-diethoxyethane
Synonyms: ethane, 2-chloro-1,1-diethoxy-
Molecular Weight (g/mol): 152.62
Molecular Formula: C6H13ClO2
InChi Key: OVXJWSYBABKZMD-UHFFFAOYSA-N
SMILES: CCOC(CCl)OCC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)