Learn More
Thermo Scientific™ Cheminert Model C2 Microbore Injector
Can be used as an injector or switching valve
Brand:  Thermo Scientific™ CZSL10PK
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11523970
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
Specifications
Specifications
10 μL