Learn More
Hanna Instruments™ Checktemp Digital Thermometers
Product Code. 15350684
Change view
Click to view available options
Diameter (Metric) Probe:
3 mm
3.6 mm
Unit Size:
Each
Length (Metric) Probe:
105 mm
110 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Probe Unit Size Length (Metric) Probe
15350684 3.6 mm Each -
13039322 3 mm Each 105 mm
15379353 3 mm Each 110 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15350684 Supplier Hanna Instruments™ Supplier No. HI98501
  • ONLINE EXCLUSIVE PRICE
    840.00 NOK
    684.61 NOK / Pack of 1
    Save 155.39 NOK
    18% Off
Estimated Shipment: 18-11-2026
Digital thermometer with stainless steel penetration probe that delivers high accuracy temperature measurements over a wide temperature range without worrying about breakage or condensation.
  • A digital thermometer with stainless steel penetration probe. It delivers high accuracy temperature measurements over a wide temperature range without worrying about breakage or condensation
  • Checktemp™ is the ideal instrument for measuring temperature according to HACCP requirements
  • ±0.2°C (±0.5°F) - Accuracy
  • Most accurate temperature tester in the world
  • CAL Check™ - Automatically verifies calibration at startup
  • °C/°F Status – User selectable
  • Large display with wide environmental temperature range and viewing angle.
  • IP 65 water resistant protection
  • HACCP Compatible - Use as a tool for control in HACCP analysis
  • AISI 316 stainless steel penetration Probe
  • 3000 hours of battery life (Continuous use)
Specifications
Certifications/Compliance AISI 316, HACCP Compatible
Diameter (Metric) Probe 3.6 mm
For Use With (Application) Education, food (storage, transportation, manufacturing, cooking), laboratory
Includes Probe
Probe Type Fixed
Resolution 0.1°C
Weight (Metric) 51.3 g
Product Type Digital Thermometer
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +150°C
Calibrated Yes
Accuracy ±0.3°C
Display Digital