Learn More
Hanna Instruments™ Checkfridge™ Remote Sensor Thermometer
Product Code. 10026121
Click to view available options
Temperature Range (Metric):
50°C to 150°C
Unit Size:
Each
Product Code. 10026121
Brand: Hanna Instruments™ HI14700
Only null left
Add to basket
Add to basket
Designed to monitor temperatures of fridges, freezers and other appliances. Hanna Instruments™ Checkfridge™ Remote Sensor Thermometer is a magnetic, continuous readout digital thermometer fitted with a stainless steel-tipped probe on a 1m cable. Unique CAL Check™ feature provides internal calibration of the device with the flick of a switch, alleviating the time preparing an ice bath.
A magnetic, continuous readout digital thermometer fitted with a stainless steel-tipped probe on a 1m cable. Unique CAL Check™ feature allows for an internal calibration of the device with the flick of a switch, thereby alleviating the time preparing an ice bath.
  • Designed to monitor temperatures of fridges, freezers and other appliances.
  • Two strong magnets are incorporated on the back of the meter that enable it to be securely fixed to any steel surface.
  • The sensor can be used for either air or product temperature monitoring.
  • A built-in calibration check device is activated at the touch of a button to ensure accurate readings at all times.
  • 1 x 1.5V AAA Battery included.
Specifications
Calibration Points 2 Points
Diameter (Metric) Probe 5 mm
Includes AAA battery
Length (Metric) Cable 1 m
Probe Type Stainless Steel General Purpose
Resolution 0.1° C
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) 50°C
Temperature Range (Metric) 50°C to 150°C
Calibrated Yes
Accuracy ±0.3°C (−20 to 90°C)
±0.5°C (outside)