Learn More
Memmert™ Chamber Modification (for Reinforced Shelves or Grids) for UN450plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN450plus Oven
Unit Size:
Each
Description
- Includes replacement of standard grids by reinforced grids
Specifications
Specifications
| For Use With (Equipment) | UN450plus Oven |
| Type | Chamber Modification |