missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceruloplasmin Mouse anti-Human, Rat, Clone: 3C8A5, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Ceruloplasmin |
|---|---|
| Clone | 3C8A5 |
| Concentration | 0.89 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857775
|
Proteintech
66156-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847775
|
Proteintech
66156-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe (II)transferrin to Fe (III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.Specifications
| Ceruloplasmin | |
| 0.89 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Ceruloplasmin, ceruloplasmin (ferroxidase), CP, CP 2, Ferroxidase | |
| CP | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3C8A5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P00450, P13635 | |
| 1356, 24268 | |
| Ceruloplasmin Fusion Protein Ag15535 | |
| Primary | |
| -20°C | |
| CP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title