missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceruloplasmin Mouse anti-Human, Rat, Clone: 3C8A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66156-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe (II)transferrin to Fe (III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.Specifications
| Ceruloplasmin | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00450, P13635 | |
| CP | |
| Ceruloplasmin Fusion Protein Ag15535 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3C8A5 | |
| Unconjugated | |
| CP | |
| Ceruloplasmin, ceruloplasmin (ferroxidase), CP, CP 2, Ferroxidase | |
| Mouse | |
| Protein G | |
| RUO | |
| 1356, 24268 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction