Learn More
Eppendorf™ Centrifuge Adapters
Product Code. p-8003599 Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 15 mL Conical Tube
1 x 7 to 15 mL Blood Collection Tube
Type:
Large Bore Adapter
Small Bore Adapter
Tube Adapter
For Use With (Equipment):
Rotor A-8-17
Rotor F-35-6-30
Unit Size:
Pack of 2
Pack of 8
Product Code. 10592774
Brand: Eppendorf™ 5427734007
Designed for use with Eppendorf Rotors and Centrifuges
  • Accessory for Eppendorf liquid handling equipment
TRUSTED_SUSTAINABILITY
Specifications
Type Large Bore Adapter
Capacity 1 x 7 to 15 mL Blood Collection Tube
Max. Tube Diameter 16 mm
Max. Tube Length 90 to 105 mm
For Use With (Equipment) Rotor F-35-6-30
Description Adapter, large bore
No. per Pack 2/Pk.