Learn More
Eppendorf™ Centrifuge Adapters
Product Code. 10638004 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1 x 15 mL Conical Tube
1 x 7 to 15 mL Blood Collection Tube
Type:
Large Bore Adapter
Small Bore Adapter
Tube Adapter
For Use With (Equipment):
Rotor A-8-17
Rotor F-35-6-30
Unit Size:
Pack of 2
Pack of 8
Product Code. Capacity Type For Use With (Equipment) unitSize
10638004 1 x 7 to 15 mL Blood Collection Tube Small Bore Adapter Rotor F-35-6-30 Pack of 2
10592774 1 x 7 to 15 mL Blood Collection Tube Large Bore Adapter Rotor F-35-6-30 Pack of 2
10789272 1 x 15 mL Conical Tube Tube Adapter Rotor A-8-17 Pack of 8
3 options
Product Code. 10638004
Brand: Eppendorf™ 5427732004
Designed for use with Eppendorf Rotors and Centrifuges
  • Accessory for Eppendorf liquid handling equipment
TRUSTED_SUSTAINABILITY
Specifications
Type Small Bore Adapter
Capacity 1 x 7 to 15 mL Blood Collection Tube
Max. Tube Diameter 16 mm
Max. Tube Length 75 to 105 mm
For Use With (Equipment) Rotor F-35-6-30
Description Adapter, small bore
No. per Pack 2/Pk.