Learn More
Eppendorf™ Centrifuge 5920 R - Refrigerated Large Centrifuge
BIOPHARMAGREENER_CHOICE
Product Code. 15546136 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
4 x 1000 mL
Includes:
Without Rotor
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Includes Voltage unitSize
15546136 4 x 1000 mL Without Rotor 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15546136 Supplier Eppendorf™ Supplier No. 5948000018
The refrigerated Eppendorf Centrifuge 5920 R offers high-throughput performance and advanced temperature management in a very compact design. With a capacity of up to 4 x 1,000 mL / 52 x 50 mL conical tubes, it can spin both plates and tubes in the same bucket for real flexibility.
  • Centrifuge lid with soft-touch lid closure.
  • Quiet operation to improve your work environment.
  • Compact footprint saves valuable bench space.
  • Automatic rotor recognition and imbalance detection for maximum operational safety.
  • Aerosol-tight Eppendorf QuickLock™ caps and lids allow fast and ergonomic lid locking.
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL.
  • Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates.
  • Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL.
  • FastTemp function for quick pre-cooling.
  • FastTemp pro™ function allows for automatic precooling based on pre-programmable time and date.
  • Continuous cooling maintains a constant temperature after the run has been completed – your samples stay cool.
  • ECO shut-off engages after 8 hours (adjustable) of non-use to reduce energy consumption and to extend compressor life.
  • Dynamic compressor control (DCC) technology for precise cooling performance.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Controller Type Keypad
Capacity 4 x 1000 mL
Max. RCF 21,194 x g
Max. Speed 13,700 rpm
Product Line Eppendorf 5920 R
Refrigerated Yes
Includes Without Rotor
Model 5920 R (EU-IVD), keypad
Noise Level <55 dB(A)
Rotor Not Included
Rotor Included No
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition
Standards EN 61010-1, EN 61010-2-020, EN 61326-1, EN 55011, CFR 47 FCC part 15 class A, EN 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-3, EN ISO 18113-1, EN ISO 15223-1, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
Timer 10 sec. to 99 hr. 59 min., Continuous Run Function, Short-spin
Dimensions (L x W x H) Exterior 74 x 71 x 40 cm (29.2 x 27.8 x 15.9 in.)
Width (English) Exterior 29.2 in.
Width (Metric) Exterior 74 cm
Height (English) Exterior 15.9 in.
Height (Metric) Exterior 40 cm
Electrical Requirements 230 V, 50/60 Hz (EU)
Voltage 230 V
Wattage 1650 W
Frequency 50/60 Hz
Product Type Benchtop Centrifuge
Display Type Large, Brightly Lit LCD