Learn More
Eppendorf™ Centrifuge 5810/5810 R
BIOPHARMAGREENER_CHOICE
Product Code. 10052674 Shop All Eppendorf Products
Change view
Click to view available options
Includes:
Rotor A-4-62 incl. adapters for 15/50 mL conical tubes
Rotor A-4-81 incl. adapters for 15/50 mL conical tubes
Rotor S-4-104 incl. adapters for 13/16 mm round-bottom tubes
Rotor S-4-104 incl. adapters for 15/50 mL conical tubes
Without rotor
Model:
Centrifuge 5810
Centrifuge 5810 (EU-IVD)
Centrifuge 5810 R
Centrifuge 5810 R (EU-IVD)
Refrigerated:
No
Yes
Electrical Requirements:
230 V, 50/60 Hz (EU)
Unit Size:
1 set
Each
Product Code. Includes Model Refrigerated Electrical Requirements unitSize
10052674 Rotor A-4-62 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 R (EU-IVD) Yes 230 V, 50/60 Hz (EU) Each
10595074 Rotor A-4-62 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 (EU-IVD) No 230 V, 50/60 Hz (EU) Each
10470975 Rotor A-4-81 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 (EU-IVD) No 230 V, 50/60 Hz (EU) Each
15127765 Rotor S-4-104 incl. adapters for 13/16 mm round-bottom tubes Centrifuge 5810 (EU-IVD) No 230 V, 50/60 Hz (EU) Each
12826213 Rotor S-4-104 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 (EU-IVD) No 230 V, 50/60 Hz (EU) Each
10000461 Without rotor Centrifuge 5810 (EU-IVD) No 230 V, 50/60 Hz (EU) Each
15805113 Rotor A-4-62 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 No 230 V, 50/60 Hz (EU) 1 set
15825113 Rotor A-4-81 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 No 230 V, 50/60 Hz (EU) 1 set
15865113 Rotor S-4-104 incl. adapters for 13/16 mm round-bottom tubes Centrifuge 5810 No 230 V, 50/60 Hz (EU) 1 set
15845113 Rotor S-4-104 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 No 230 V, 50/60 Hz (EU) 1 set
15885103 Without rotor Centrifuge 5810 No 230 V, 50/60 Hz (EU) Each
10012724 Rotor A-4-81 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 R (EU-IVD) Yes 230 V, 50/60 Hz (EU) Each
15137765 Rotor S-4-104 incl. adapters for 13/16 mm round-bottom tubes Centrifuge 5810 R (EU-IVD) Yes 230 V, 50/60 Hz (EU) Each
12836213 Rotor S-4-104 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 R (EU-IVD) Yes 230 V, 50/60 Hz (EU) Each
10460713 Without rotor Centrifuge 5810 R (EU-IVD) Yes 230 V, 50/60 Hz (EU) Each
15805123 Rotor A-4-62 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 R Yes 230 V, 50/60 Hz (EU) 1 set
15825123 Rotor A-4-81 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 R Yes 230 V, 50/60 Hz (EU) 1 set
15865123 Rotor S-4-104 incl. adapters for 13/16 mm round-bottom tubes Centrifuge 5810 R Yes 230 V, 50/60 Hz (EU) 1 set
15845123 Rotor S-4-104 incl. adapters for 15/50 mL conical tubes Centrifuge 5810 R Yes 230 V, 50/60 Hz (EU) 1 set
15885113 Without rotor Centrifuge 5810 R Yes 230 V, 50/60 Hz (EU) Each
20 options
Product Code. 10052674
Brand: Eppendorf™ 5811000320
The Eppendorf Centrifuge 5810/5810 R is the perfect day-to-day centrifuge for laboratories with medium-throughput requirements. It has an extremely compact footprint yet still offers the real versatility of being able to take both tube and plate formats.
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 750 mL
  • Plate rotors for centrifugation of all types of MTP, PCR, cell culture, or Deepwell Plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 mL to 85 mL
  • High centrifugation speed of up to 20,913 × g (14,000 rpm)
  • Aerosol-tight Eppendorf QuickLock™ caps and lids for easy, one-hand operation
  • Centrifuge lid with soft-touch lid closure
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve your work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Controller Type Keypad
    Capacity 4 x 750 mL
    Max. RCF 20,913 x g
    Max. Speed 14,000 rpm
    Refrigerated Yes
    Display Large, brightly lit LCD
    Includes Rotor A-4-62 incl. adapters for 15/50 mL conical tubes
    Maximum Speed Range 200 to 14,000 rpm
    Model Centrifuge 5810 R (EU-IVD)
    Noise Level <56 dB(A)
    Run Time 1 to 99 min.
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition
    Standards EN 61010-1, EN 61010-2-020, EN 61000-6-1, EN 61000-3-2, EN 61000-3-3, EN 61326-1, EN 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-3, EN ISO 18113-1, EN ISO 15223-1, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
    Temperature Range 15°C to 35°C
    Timer 1 to 99 min., with continuous run function, short-spin
    Type Benchtop Centrifuges
    Width (English) Exterior 27.6 in.
    Width (Metric) Exterior 70.0 cm
    Height (English) Exterior 13.6 in.
    Height (Metric) Exterior 34.5 cm
    Depth (Metric) 608 mm
    Net Weight 99 kg
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Voltage 230 V
    Wattage 1650 W
    Show More Show Less
    The Centrifuge 5810/5810 R is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose. This product complies with the requirements of the EU In Vitro Diagnostics Regulation (IVDR) 2017/746. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.