Learn More
Eppendorf™ Centrifuge 5804/5804 R
BIOPHARMAGREENER_CHOICE
Product Code. 10246074 Shop All Eppendorf Products
Change view
Click to view available options
Includes:
Rotor A-4-44 incl. adapters for 15/50 mL conical tubes
Rotor S-4-72 incl. adapters for 15/50 mL conical tubes
Without rotor
Electrical Requirements:
230 V, 50/60 Hz (EU)
Model:
Centrifuge 5804
Centrifuge 5804 (EU-IVD)
Centrifuge 5804 R
Centrifuge 5804 R (EU-IVD)
Unit Size:
1 set
Each
Product Code. Includes Electrical Requirements Model unitSize
10246074 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R (EU-IVD) Each
10022724 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 (EU-IVD) Each
12806213 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 (EU-IVD) Each
10344701 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 (EU-IVD) Each
15885093 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 1 set
15805103 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 1 set
15865093 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 Each
12816213 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R (EU-IVD) Each
10334751 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 R (EU-IVD) Each
15845103 Rotor A-4-44 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R 1 set
15865103 Rotor S-4-72 incl. adapters for 15/50 mL conical tubes 230 V, 50/60 Hz (EU) Centrifuge 5804 R 1 set
15825103 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5804 R Each
12 options
Product Code. 10246074
Brand: Eppendorf™ 5805000327
The high-speed Eppendorf Centrifuge 5804/5804 R is a medium capacity centrifuge, ideal for molecular applications in tubes up to 250 mL. It also offers supreme versatility with the capability of taking swing-bucket and fixed-angle rotors, as well as deepwell plates.
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 250 mL
  • Plate rotor for centrifugation of all types of MTP, PCR, cell culture, or Deepwell Plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 mL to 85 mL
  • High centrifugation speed of up to 20,913 × g (14,000 rpm)
  • Centrifuge lid with soft-touch lid closure
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve your work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Controller Type Keypad
    Capacity 4 x 250 mL
    Max. RCF 20,913 x g
    Refrigerated Yes
    Display Large, brightly lit LCD
    Includes Rotor A-4-44 incl. adapters for 15/50 mL conical tubes
    Maximum Speed Range 200 to 14,000 rpm
    Model Centrifuge 5804 R (EU-IVD)
    Noise Level <58 dB(A)
    Run Time 1 to 99 min.
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition
    Standards EN 61010-1, EN 61010-2-020, EN 61000-6-1, EN 61000-3-2, EN 61000-3-3, EN 61326-1, EN 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-3, EN ISO 18113-1, EN ISO 15223-1, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
    Temperature Range 15°C to 35°C
    Timer 1 to 99 min., with continuous run function, short-spin
    Type Benchtop Centrifuges
    Width (English) Exterior 25.0 in.
    Width (Metric) Exterior 63.4 cm
    Height (English) Exterior 13.5 in.
    Height (Metric) Exterior 34.2 cm
    Depth (Metric) 550 mm
    Net Weight 80 kg
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Voltage 230 V
    Wattage 1650 W