Learn More
Eppendorf™ Centrifuge 5702/5702 R/5702 RH
BIOPHARMAGREENER_CHOICE
Product Code. 15865083 Shop All Eppendorf Products
Change view
Click to view available options
Includes:
Rotor A-4-38 incl. adapters for 13/16 mm blood collection tubes, 2 sets of 2 adapters
Rotor A-4-38 incl. adapters for 15/50 mL conical tubes, 2 sets of 2 adapters
Rotor A-4-38 incl. adapters for round-bottom tubes 4 - 10 mL and adapters for round-bottom tubes 2.6 - 7 mL, 2 sets of 2 adapters
Without rotor
Electrical Requirements:
230 V, 50/60 Hz (EU)
Model:
Centrifuge 5702
Centrifuge 5702 (EU-IVD)
Centrifuge 5702 R
Centrifuge 5702 R (EU-IVD)
Centrifuge 5702 RH
Centrifuge 5702 RH (EU-IVD)
Unit Size:
1 set
Each
Product Code. Includes Electrical Requirements Model unitSize
15865083 Rotor A-4-38 incl. adapters for 15/50 mL conical tubes, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 1 set
10746805 Rotor A-4-38 incl. adapters for 15/50 mL conical tubes, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 (EU-IVD) Each
17322831 Rotor A-4-38 incl. adapters for round-bottom tubes 4 - 10 mL and adapters for round-bottom tubes 2.6 - 7 mL, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 (EU-IVD) 1 set
10469451 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5702 (EU-IVD) Each
17332831 Rotor A-4-38 incl. adapters for round-bottom tubes 4 - 10 mL and adapters for round-bottom tubes 2.6 - 7 mL, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 1 set
15845083 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5702 Each
10194444 Rotor A-4-38 incl. adapters for 13/16 mm blood collection tubes, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 R (EU-IVD) Each
12689355 Rotor A-4-38 incl. adapters for 15/50 mL conical tubes, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 R (EU-IVD) Each
10772963 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5702 R Each
15805093 Rotor A-4-38 incl. adapters for 13/16 mm blood collection tubes, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 R 1 set
15825093 Rotor A-4-38 incl. adapters for 15/50 mL conical tubes, 2 sets of 2 adapters 230 V, 50/60 Hz (EU) Centrifuge 5702 R 1 set
15885083 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5702 R Each
10485762 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5702 RH (EU-IVD) Each
15845093 Without rotor 230 V, 50/60 Hz (EU) Centrifuge 5702 RH Each
14 options
Product Code. 15865083
Brand: Eppendorf™ 5702000320
The range of Eppendorf 5702 general purpose, low-speed centrifuges has been designed specifically to meet the needs of mid-throughput clinical research and cell culture laboratories. These centrifuges are very compact, so that they fit easily on the lab bench, as well as extremely quiet.
  • Max. speed: 3,000 × g (4,400 rpm)
  • Very compact footprint to fit on every lab bench
  • Very quiet operation to benefit your environment
  • Low access height for simple loading and unloading of samples
  • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
  • 'At set RPM' function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
  • Key lock to prevent unintentional adjustments
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Electronic imbalance detection for maximum safety
  • TRUSTED_SUSTAINABILITY
    Specifications
    Includes Rotor A-4-38 incl. adapters for 15/50 mL conical tubes, 2 sets of 2 adapters
    Max. RCF 3,000 x g
    Max. Speed 4,400 rpm
    Refrigerated No
    Noise Level <52 dB(A) with Rotor A-4-38
    Dimensions (L x W x H) Exterior 320 x 400 x 240 mm
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Display LCD
    Height (English) Exterior 9.6 in.
    Height (Metric) Exterior 24 cm
    Timer 0 to 99 min., with continuous run function
    Width (English) Exterior 12.6 in.
    Width (Metric) Exterior 32 cm
    Wattage 200 W
    Controller Type Rotary Knob
    Model Centrifuge 5702
    Ventilated Yes
    Rotor Included Yes
    Frequency 50/60 Hz
    Voltage 230 V
    Show More Show Less
    The Centrifuge 5702/5702 R/5702 RH is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose. This product complies with the requirements of the EU In Vitro Diagnostics Regulation (IVDR) 2017/746. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.