Learn More
Eppendorf™ Centrifuge 5425/5425 R
BIOPHARMAGREENER_CHOICE
Product Code. 15891635 Shop All Eppendorf Products
Change view
Click to view available options
Electrical Requirements:
230V/50–60Hz (EU)
230V/50–60Hz (GB)
Model:
Centrifuge 5425
Centrifuge 5425 (EU-IVD)
Centrifuge 5425 R
Centrifuge 5425 R (EU-IVD)
Refrigerated:
No
Yes
Rotor Included:
FA-24x2
No
Yes
Unit Size:
1 set
Each
Product Code. Electrical Requirements Model Refrigerated Rotor Included unitSize
15891635 230V/50–60Hz (EU) Centrifuge 5425 (EU-IVD) No No Each
15831645 230V/50–60Hz (EU) Centrifuge 5425 (EU-IVD) No Yes Each
15841645 230V/50–60Hz (GB) Centrifuge 5425 (EU-IVD) No Yes Each
15851635 230V/50–60Hz (EU) Centrifuge 5425 (EU-IVD) No Yes Each
15861635 230V/50–60Hz (GB) Centrifuge 5425 (EU-IVD) No Yes Each
15871645 230V/50–60Hz (EU) Centrifuge 5425 (EU-IVD) No No Each
15811645 230V/50–60Hz (EU) Centrifuge 5425 No No Each
15851645 230V/50–60Hz (EU) Centrifuge 5425 No FA-24x2 Each
15861645 230V/50–60Hz (GB) Centrifuge 5425 No Yes Each
15871635 230V/50–60Hz (EU) Centrifuge 5425 No Yes Each
15881635 230V/50–60Hz (GB) Centrifuge 5425 No Yes Each
15891645 230V/50–60Hz (EU) Centrifuge 5425 No No Each
16855100 230V/50–60Hz (EU) Centrifuge 5425 R Yes Yes 1 set
16895100 230V/50–60Hz (EU) Centrifuge 5425 R Yes No Each
16835110 230V/50–60Hz (EU) Centrifuge 5425 R Yes Yes 1 set
16875110 230V/50–60Hz (EU) Centrifuge 5425 R Yes No Each
16835100 230V/50–60Hz (EU) Centrifuge 5425 R (EU-IVD) Yes Yes 1 set
16875100 230V/50–60Hz (EU) Centrifuge 5425 R (EU-IVD) Yes No Each
16815110 230V/50–60Hz (EU) Centrifuge 5425 R (EU-IVD) Yes Yes 1 set
16855110 230V/50–60Hz (EU) Centrifuge 5425 R (EU-IVD) Yes No Each
16672832 230V/50–60Hz (EU) Centrifuge 5425 No Yes Each
17474152 230V/50–60Hz (EU) Centrifuge 5425 (EU-IVD) No Yes 1 set
22 options
Product Code. 15891635
Brand: Eppendorf™ 5405000212
Eppendorf’s newest 24-place microcentrifuges, Centrifuge 5425 and 5425 R, feature ergonomic operation and a large selection of rotors with a maximum speed of up to 21,300 x g, making them perfectly suited for all modern molecular biological applications in Eppendorf and PCR tubes from 0.2 to 5 mL.
For processing temperature-sensitive samples, the refrigerated Centrifuge 5425 R provides precise temperature control with its advanced cooling system.
  • Aerosol-tight Eppendorf QuickLock™ lids for simple and safe handling
  • Outstanding software features (e.g. adjustable acceleration and braking ramps, 3 program buttons (only Centrifuge 5425), short function without continuous pressing of the key)
  • Extended rotor program (e.g. 5.0 mL rotor, swing-bucket rotor for PCR strips)
  • OptiBowl™ design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
  • Max. rotor capacity: 10 × 5.0 mL vessels
  • Max. speed: 21,300 × g (15,060 rpm)
TRUSTED_SUSTAINABILITY
Specifications
Capacity 24 x 1.5/2 mL
Max. Speed 15,000 rpm
Max. RCF 21,300 x g
Display Large, brightly lit LCD
Height (English) Exterior 9.5 in.
Height (Metric) Exterior 24 cm
Noise Level Average of 45 dB(A) in the first 12 minutes and < 51 dB(A) thereafter with Rotor FA-24 x 2
Timer 10 s to 9:59 h, with continuous run function
Width (English) Exterior 9.5 in.
Width (Metric) Exterior 24 cm
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Type Microcentrifuges
Includes Without rotor
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
Ventilated Yes
Electrical Requirements 230V/50–60Hz (EU)
Controller Type Keypad
Depth (Metric) 39 cm
Model Centrifuge 5425 (EU-IVD)
Wattage 280 W
Rotor Included No
Refrigerated No
Run Time 10 sec. to 9 hr. 59 min.
Standards EN ISO 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-1, EN ISO 18113-3, EN ISO 15223-1, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020, EN 55011, EN 61326-1, EN 50581, IEC 61010-2-101, UL 61010-1, UL 61010-2-020, CAN/CSA-C22.2 No. 61010-1, CAN/CSA-C22.2 No. 61010-2-020, 47 CFR FCC Part 15 (refer to manual for more details)
Net Weight 15.6 kg
Frequency 50/60 Hz
Voltage 230 V
Show More Show Less
The Centrifuge 5425 is designed for separating liquid substance mixtures with different densities, in particular, for processing and analyzing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose. This centrifuge including components is an in-vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.