Learn More
CENPF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28568-1-AP-20UL
 View more versions of this product
Product Code. 17669636
  • 1710.33 NOK / 20µL
Description
CENPF Polyclonal antibody apecifically detetects CENPF antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.
Specifications
Specifications
CENPF
Polyclonal
Unconjugated
NM_016343
Rabbit
Antigen affinity purification
Primary
Human
Liquid
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
700 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
CENPF
CENPF fusion protein Ag29665
20 μL
1063
Store at -20°C. Stable for one year after shipment.