Learn More
CENPF Rabbit anti-Human, Polyclonal, Proteintech
Product Code. p-7225923 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17669636 20 μL 20µL
17659636 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17669636 Supplier Proteintech Supplier No. 285681AP20UL
Rabbit Polyclonal Antibody
CENPF Polyclonal antibody apecifically detetects CENPF antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.
Specifications
Antigen CENPF
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 700 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. NM_016343
Gene Symbols CENPF
Host Species Rabbit
Immunogen CENPF fusion protein Ag29665
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1063
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG