Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11327, 0.2μm
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11327, 0.2μm are recommended for a broad range of applications including liquid filtering for sample preparation.
Brand: Sartorius Stedim Biotech™ 11327--25------N
Additional Details : Weight : 0.08100kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Cellulose nitrate (CN), mixed cellulose ester (MCE), membrane filters are composed of cellulose nitrate and a small content of cellulose acetate.
- Recommended for a broad range of applications such as the filtration of aqueous solutions, air/gas filtration, or sample preparation.
- Suitable for sterile filtration, analytical sample preparation, clarification, particle analysis.
- Cellulose nitrate filters, pore size 0.2 μm can be used for the analysis of filterability of beer (membrane filter test according to Esser, in “MEBAK – Brautechnische Analysemethoden – Würze, Bier, Mischgetränke – Methodensammlung der Mitteleuropäischen Brautechnischen Analysenkommission“ 2012, Chapter 2.20.1)
- Sterilizable by autoclave, gamma irradiation, or ETO.
- Non-sterile, bulk-packed.
Specifications
White | |
0.2 μm | |
11327 | |
100/pk. | |
25 mm |
Cellulose Nitrate, Mixed Cellulose Ester | |
Non-sterile | |
Hydrophilic | |
130 μm |
Safety and Handling
- Danger
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose