Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11327, 0.2μm
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11327, 0.2μm are recommended for a broad range of applications including liquid filtering for sample preparation.
1245.00 NOK - 2215.00 NOK
Specifications
Color | White |
---|---|
Material | Cellulose Nitrate, Mixed Cellulose Ester |
Pore Size | 0.2 μm |
Sterility | Non-sterile |
Grade | 11327 |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15902460
|
Sartorius Stedim Biotech™
11327--25------N |
25 mm |
1245.00 NOK
Pack of 100 |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912460
|
Sartorius Stedim Biotech™
11327--47------N |
47 mm |
2215.00 NOK
Pack of 100 |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Cellulose nitrate (CN), mixed cellulose ester (MCE), membrane filters are composed of cellulose nitrate and a small content of cellulose acetate.
- Recommended for a broad range of applications such as the filtration of aqueous solutions, air/gas filtration, or sample preparation.
- Suitable for sterile filtration, analytical sample preparation, clarification, particle analysis.
- Cellulose nitrate filters, pore size 0.2 μm can be used for the analysis of filterability of beer (membrane filter test according to Esser, in “MEBAK – Brautechnische Analysemethoden – Würze, Bier, Mischgetränke – Methodensammlung der Mitteleuropäischen Brautechnischen Analysenkommission“ 2012, Chapter 2.20.1)
- Sterilizable by autoclave, gamma irradiation, or ETO.
- Non-sterile, bulk-packed.
Specifications
White | |
0.2 μm | |
11327 | |
100/pk. |
Cellulose Nitrate, Mixed Cellulose Ester | |
Non-sterile | |
Hydrophilic | |
130 μm |