Learn More
Biodesign™ CellMicroSieves™
Product Code. 12924267
Change view
Click to view available options
Pore Size:
20 μm
25 μm
50 μm
70 μm
100 μm
Format:
Circle
Roll
Packungsgröße:
Each
Pack of 10
Artikelnummer. Pore Size Format unitSize
12924267 70 μm Roll Each
12924277 100 μm Circle Pack of 10
12974277 20 μm Circle Pack of 10
12964267 20 μm Roll Each
12964277 25 μm Circle Pack of 10
12944277 50 μm Circle Pack of 10
12934277 70 μm Circle Pack of 10
7 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12924267
Marke: Biodesign™ N70R
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory. AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300mL capacity are autoclavable and reusable. Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.
  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.
Spezifikation
Pore Size 70 μm
Type Cell Micro Sieve
Format Roll