Learn More
Molecular Biology Grade Water Tested to USP Sterile water, Corning™
BIOPHARMA
Product Code. p-6983003
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
6 x 1 L
Unit Size:
Pack of 6
Product Code. 15383581
Brand: Corning Media Tech 46000CI
Used in RNase, DNase and Protease free applications. Corning™ Molecular Biology Grade Water Tested to USP Sterile water is used for Buffer preparation, DNA/RNA purification, DNA sequencing and libraries, recombinant DNA, cloning, gene mapping.
Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 6 x 100 mL