missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK4 Mouse anti-Human, Unconjugated, Clone: 1G2C12, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CDK4 |
---|---|
Clone | 1G2C12 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889265
|
Proteintech
66950-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879265
|
Proteintech
66950-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CDK4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK4. This antibody recognizes human, mouse, rat antigen. The CDK4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CDK4 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1019 | |
Primary |
1G2C12 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |