Learn More
CDK4 Mouse anti-Human, Unconjugated, Clone: 1G2C12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66950-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889265
  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
Description
The CDK4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK4. This antibody recognizes human, mouse, rat antigen. The CDK4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CDK4
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry
1G2C12
Unconjugated
20 μL
1019
IgG1