missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC2L6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | CDC2L6 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16822144
|
Proteintech
13761-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16812144
|
Proteintech
13761-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC2L6 is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.Specifications
| CDC2L6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8BWD8, Q9BWU1 | |
| 23097, 309804, 78334 | |
| CDC2L6 Fusion Protein Ag4731 | |
| Primary | |
| -20°C | |
| CDK19 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CDC2 related protein kinase 6, CDC2L6, CDK11, CDK19, Cyclin dependent kinase 11, Death preventing kinase, KIAA1028 | |
| Cdk19 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title