Learn More
CDC2L6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13761-1-AP-20UL
 View more versions of this product
Product Code. 16822144
  • 1710.33 NOK / 20µL
Description
CDC2L6 is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.
Specifications
Specifications
CDC2L6
Polyclonal
Unconjugated
CDK19
CDC2 related protein kinase 6, CDC2L6, CDK11, CDK19, Cyclin dependent kinase 11, Death preventing kinase, KIAA1028
Rabbit
Antigen Affinity Chromatography
RUO
23097, 309804, 78334
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BWD8, Q9BWU1
Cdk19
CDC2L6 Fusion Protein Ag4731
20 μL
Primary
Mouse, Rat, Human