Learn More
IBA Lifesciences CD9 Fab-TACS™ Exosome Agarose Column Starter Kit
Shop All IBA Lifesciences ProductsDescription
- Fab-TACS™ is a traceless affinity exosome isolation approach based on our Strep-tag™ technology
- CD9 Fab-TACS™ Exosome Agarose Column Starter Kit allows label-free isolation of human CD9+ exosomes from plasma, serum and cell culture supernatants
- CD9 is a member of the tetraspanin family present on the surface of exosomes
- Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
- Exosomes adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target particles are efficiently washed away
- In a final step, the addition of biotin causes the elution of the exosomes and the Fab-Streps. After elution, the Fab-Streps self-dissociate from the exosome surface due to their low affinity
- The label-free, fully functional exosomes are now ready for further downstream applications.
Spécification
Spécification
| Includes | Strep-Tactin™ TACS Agarose columns, 0.3 mL (2 x), CD9 Fab-Strep (human), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter |
| Target Species | Human |
For Research Use Only.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?