Learn More
CD8a Mouse, anti-Human, PerCP-Cyanine5.5, Clone: RPA-T8, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60022PS.1
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17108551
  • 2267.80 NOK / 25 tests
Estimated Shipment: 13-10-2023
Description
  • The RPA-T8 antibody reacts with CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
  • CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
  • CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
  • It functions to strengthen the association between the TCR and MHC I-antigen complex
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD8+ T Cell Isolation Kit (Catalog #19663), EasySep™ Human CD8+ T Cell Enrichment Kit (Catalog #19053), EasySep™ Human CD8 Positive Selection Kit II (Catalog #17853), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851), EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883), and EasySep™ HLA T Cell Enrichment Kit (Catalog #19051HLA)
Specifications
Specifications
CD8a
Monoclonal
PerCP-Cyanine5.5
25 Tests
Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human
T Cells, T Cells, CD8+
CyTOF, Flow Cytometry, Functional Assay, Immunohistochemistry
RPA-T8
Mouse
Primary
IgG1 κ