missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Mouse, anti-Human, PerCP-Cyanine5.5, Clone: RPA-T8, STEMCELL Technologies
Mouse Monoclonal Antibody
POA - 4968.65 NOK
Specifications
| Antigen | CD8a |
|---|---|
| Clone | RPA-T8 |
| Applications | CyTOF, Flow Cytometry, Functional Assay, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | PerCP-Cyanine5.5 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17108551
|
Stemcell Technologies
60022PS.1 |
25 Tests | |||||||
17198541
|
Stemcell Technologies
60022PS |
100 Tests |
4968.65 NOK
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The RPA-T8 antibody reacts with CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
- CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
- CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
- It functions to strengthen the association between the TCR and MHC I-antigen complex
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD8+ T Cell Isolation Kit (Catalog #19663), EasySep™ Human CD8+ T Cell Enrichment Kit (Catalog #19053), EasySep™ Human CD8 Positive Selection Kit II (Catalog #17853), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851), EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883), and EasySep™ HLA T Cell Enrichment Kit (Catalog #19051HLA)
Specifications
| CD8a | |
| CyTOF, Flow Cytometry, Functional Assay, Immunohistochemistry | |
| PerCP-Cyanine5.5 | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Primary |
| RPA-T8 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title