Learn More
CD8a Mouse, anti-Human, PerCP-Cyanine5.5, Clone: RPA-T8, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60022PS
Encompass_Preferred
 View more versions of this product
Product Code. 17198541
  • 4968.65 NOK / 100 tests
Description
  • The RPA-T8 antibody reacts with CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
  • CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
  • CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
  • It functions to strengthen the association between the TCR and MHC I-antigen complex
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD8+ T Cell Isolation Kit (Catalog #19663), EasySep™ Human CD8+ T Cell Enrichment Kit (Catalog #19053), EasySep™ Human CD8 Positive Selection Kit II (Catalog #17853), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851), EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883), and EasySep™ HLA T Cell Enrichment Kit (Catalog #19051HLA)
Specifications
Specifications
CD8a
Monoclonal
PerCP-Cyanine5.5
100 Tests
Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human
T Cells, T Cells, CD8+
CyTOF, Flow Cytometry, Functional Assay, Immunohistochemistry
RPA-T8
Mouse
Primary
IgG1 κ
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.