Learn More
Stemcell Technologies CD71 (Transferrin Receptor) Mouse, anti-Human, APC, Clone: OKT9, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60106AZ.1
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17109051
  • 1971.88 NOK / 25 tests
Estimated Shipment: 20-06-2023
to see stock.
Description
Description
  • The OKT9 antibody reacts with CD71 (transferrin receptor), an ∼180 - 190 kDa disulfide-bonded homodimer and type 2 transmembrane glycoprotein expressed at high levels on a broad range of actively proliferating cells, as well as reticulocytes, monocytes, macrophages and marrow stromal cells
  • Surface levels are low on resting leukocytes but are upregulated on lymphocytes, monocytes and macrophages following antigen or mitogen stimulation
  • Expression is lost during differentiation of reticulocytes into mature erythrocytes
  • CD71 mediates uptake of transferrin–iron complexes through receptor-mediated endocytosis and recycling of the apotransferrin–receptor complex to the cell surface
  • It has also served as a target for drug delivery
  • The OKT9 antibody binds an epitope that reportedly overlaps with that of antibody clone CY1G4 but is distinct from the ligand binding site for transferrin
Specifications
Specifications
CD71 (Transferrin Receptor)
Monoclonal
APC
25 Tests
Human
Erythroid Cells
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OKT9
Mouse
Primary
IgG1 κ