missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD70 Monoclonal Antibody (OTI7B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813205
This item is not returnable.
View return policy
Description
CD70 Monoclonal antibody specifically detects CD70 in Human samples. It is validated for Flow CytometrySpecifications
| CD70 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P32970 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 970 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry | |
| OTI7B2 | |
| Unconjugated | |
| Cd70 | |
| CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7 | |
| Full length human recombinant protein of human CD70 (NP_001243) produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction