missing translation for 'onlineSavingsMsg'
Learn More

CD70 Monoclonal Antibody (OTI7B2), TrueMAB™, OriGene

Product Code. p-7225521
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17517934 30 μL 30µL
17597924 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17517934

Brand: Origene Technologies TA813205S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD70 Monoclonal antibody specifically detects CD70 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD70
Applications Flow Cytometry
Classification Monoclonal
Clone OTI7B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Cd70
Gene Accession No. P32970
Gene Alias CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7
Host Species Mouse
Immunogen Full length human recombinant protein of human CD70 (NP_001243) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 970
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.