Learn More
Novus Biologicals™ CD5 Antibody Pair [HRP]
Product Code. 18634770 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18634770
Brand: Novus Biologicals™ NBP2795765Plates
Polyclonal/Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD5.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 12.5 pg/mL
Assay Sensitivity 12.5-800 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD5.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Gene ID (Entrez) 921
Gene Symbol CD5
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 140 ng of recombinant mouse CD5.
  • Capture Antibody: 1 mg/mL of rabbit anti-mouse CD5 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-CD5 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific
    Storage Requirements Storage is content dependent.
    Target CD5
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 800 pg/mL is recommended.