missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD38 Antibody Pair [HRP]

Product Code. 18284008 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. Quantity unitSize
18284008 15 Plates Pack of 15
18638460 5 Plates Pack of 5
2 options
This item is not returnable. View return policy

Product Code. 18284008

Brand: Novus Biologicals™ NBP27930115PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human CD38.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 93.75 pg/mL
Assay Sensitivity 39.06-2500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human CD38.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Gene ID (Entrez) 952
Gene Symbol CD38
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.16 mg/mL rabbit anti-CD38 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-protector, pH 7.4).
  • Capture Antibody: 1 mg/mL of rabbit anti-CD38 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 135 ng of recombinant CD38.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target CD38
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2.5 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.