missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15426-1-AP-150UL
This item is not returnable.
View return policy
Description
CD300g receptor which may mediate L-selectin-dependent lymphocyte rollings. Binds SELL in a calcium dependent manner. Binds lymphocyte.Specifications
| CD300LG | |
| Polyclonal | |
| Unconjugated | |
| CD300LG | |
| CD300LG, CLM 9, CLM9, CMRF35 like molecule 9, NEPMUCIN, TREM 4, TREM4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146894 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UXG3 | |
| CD300LG | |
| CD300LG Fusion Protein Ag8075 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction