Learn More
IBA Lifesciences CD3 Fab-TACS™ Agarose Column Starter Kit
Label-free isolation of CD3+ T cells
1535.00 NOK - 2640.00 NOK
Specifications
Content And Storage 2°C to 8°C
View More Specs
Products 2
Product Code Brand Target Species Price Quantity & Availability  
Product Code Brand Target Species Price Quantity & Availability  
17217740
View Documents Promotion Details
IBA Lifesciences
6-3201-002
Human This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17277740
View Documents Promotion Details
IBA Lifesciences
6-3304-002
Mouse This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology
  • CD3 is a co-receptor that associates with the T cell receptor and plays a role in T cell activation
  • Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation
  • Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target cells are efficiently washed away
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps
  • After elution, the Fab-Streps self-dissociate from the cell surfaces due to their low affinity
  • The label-free authentic target cells are now ready for further downstream applications
Specifications
Specifications
2°C to 8°C
Documents
Product Certifications
For Research Use Only.