Learn More
Novus Biologicals™ CD200R1L Antibody Pair [HRP]
Product Code. 18238917 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18238917
Brand: Novus Biologicals™ NBP27966115PLATES
Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD200R1L.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 125 pg/mL
Assay Sensitivity 125-8000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD200R1L.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa
Gene ID (Entrez) 344807
Gene Symbol CD200R1L
Host Species Mouse/Rabbit
Kit Contents
  • Detection Antibody: 0.25 mg/mL mouse anti-CD200R1L monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-CD200R1L monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 80 ng of recombinant CD200R1L.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target CD200R1L
    Technique 1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 8 ng/mL is recommended.