Learn More
CD134 (OX40) Monoclonal Antibody (OX-86), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-1341-050
Additional Details : Weight : 0.06000kg
Description
The OX-86 MAb reacts with mouse CD134, also known as OX40. A member of the TNF receptor superfamily, CD134 is a 50 kDa type I membrane glycoprotein expressed by activated mouse T lymphocytes. Rat CD134 was initially identified as an activation marker only on activated CD4+ T cells. In contrast, mouse CD134 is expressed by both activated CD4+ and CD8+ T cells. The interaction of CD134 with CD252 (OX40 ligand) has been implicated in T cell-dependent humoral responses, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool, and regulation of tolerance in the CD4+ T cell compartment.
CD134 (OX40) Monoclonal antibody specifically detects CD134 (OX40) in Mouse samples. It is validated for Flow Cytometry, Functional AssaySpecifications
CD134 (OX40) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P47741 | |
Rat | |
50 mg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Functional Assay | |
OX-86 | |
Functional Grade | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; TNFRSF4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; TXGP1L | |
Affinity chromatography | |
RUO | |
22163 | |
4° C | |
Liquid |