missing translation for 'onlineSavingsMsg'
Learn More

CCDC41 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26013-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894535

  • 4593.76 NOK / 150µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Component of the distal appendage region of the centriole involved in the initiation of primary cilium assembly. May collaborate with IFT20 in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium during the initiation of primary cilium assembly.
Specifications

Specifications

CCDC41
Polyclonal
Unconjugated
CEP83
CCDC41, CEP83, NY REN 58
Rabbit
Antigen Affinity Chromatography
RUO
51134
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y592
CEP83
CCDC41 Fusion Protein Ag22826
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers