missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CCDC41 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16804545
|
Proteintech
26013-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16894535
|
Proteintech
26013-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of the distal appendage region of the centriole involved in the initiation of primary cilium assembly. May collaborate with IFT20 in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium during the initiation of primary cilium assembly.Specifications
CCDC41 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9Y592 | |
51134 | |
CCDC41 Fusion Protein Ag22826 | |
Primary | |
-20°C | |
CEP83 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCDC41, CEP83, NY REN 58 | |
CEP83 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |