missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26013-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Component of the distal appendage region of the centriole involved in the initiation of primary cilium assembly. May collaborate with IFT20 in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium during the initiation of primary cilium assembly.Specifications
CCDC41 | |
Polyclonal | |
Unconjugated | |
CEP83 | |
CCDC41, CEP83, NY REN 58 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51134 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y592 | |
CEP83 | |
CCDC41 Fusion Protein Ag22826 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |