missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC111/PRIMPOL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29824-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CCDC111/PRIMPOL Polyclonal antibody apecifically detetects CCDC111/PRIMPOL antigen in Human Samples. It is validated for Western Blot, ELISA.Specifica
CCDC111/PRIMPOL | |
Polyclonal | |
Unconjugated | |
BC064600 | |
Rabbit | |
150 μL | |
201973 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |
Western Blot, ELISA | |
650 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
CCDC111 | |
Antigen affinity purification | |
Primary | |
Human | |
Liquid |