missing translation for 'onlineSavingsMsg'
Learn More

CCDC111/PRIMPOL Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7225838 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17609706 20 μL 20µL
17699696 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17609706

Brand: Proteintech 298241AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC111/PRIMPOL Polyclonal antibody apecifically detetects CCDC111/PRIMPOL antigen in Human Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CCDC111/PRIMPOL
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 650 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC064600
Gene Symbols CCDC111
Host Species Rabbit
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 201973
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.