missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC2D2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22293-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to CC2D2A.
This gene encodes a coiled-coil and calcium binding domain protein that appears to play a critical role in cilia formation. Mutations in this gene cause Meckel syndrome type 6, as well as Joubert syndrome type 9. Alternative splicing results in multiple transcript variants.Specifications
CC2D2A | |
Polyclonal | |
Unconjugated | |
CC2D2A | |
CC2D2A, JBTS9, KIAA1345, MKS6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231214, 57545 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CFW7, Q9P2K1 | |
CC2D2A | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |