missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC2D2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CC2D2A |
---|---|
Concentration | 0.54 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891835
|
Proteintech
22293-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16881835
|
Proteintech
22293-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The antibody is specific to CC2D2A.
This gene encodes a coiled-coil and calcium binding domain protein that appears to play a critical role in cilia formation. Mutations in this gene cause Meckel syndrome type 6, as well as Joubert syndrome type 9. Alternative splicing results in multiple transcript variants.Specifications
CC2D2A | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q8CFW7, Q9P2K1 | |
231214, 57545 | |
Peptide | |
Primary | |
-20°C | |
CC2D2A |
0.54 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CC2D2A, JBTS9, KIAA1345, MKS6 | |
CC2D2A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |