missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC2D1A Antibody (SR1124), Novus Biologicals™
Rabbit Monoclonal Antibody
3050.00 NOK - 4050.00 NOK
Specifications
| Antigen | CC2D1A |
|---|---|
| Clone | SR1124 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653215
|
Novus Biologicals
NBP3-22397-100ul |
100 μg |
4050.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654475
|
Novus Biologicals
NBP3-22397-50ul |
50 μg |
3050.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CC2D1A Monoclonal antibody specifically detects CC2D1A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| CC2D1A | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Akt kinase-interacting protein 1, coiled-coil and C2 domain containing 1A, coiled-coil and C2 domain-containing protein 1A, Five repressor element under dual repression-binding protein 1, FLJ20241, FLJ41160, FRE under dual repression-binding protein 1, Freud-1, mental retardation, nonsyndromic, autosomal recessive, 3, MRT3Freud-1/Aki1, Putative NF-kappa-B-activating protein 023N, putative NFkB activating protein | |
| A synthesized peptide derived from human CC2D1A (Uniprot #: Q6P1N0) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1124 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 54862 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title