missing translation for 'onlineSavingsMsg'
Learn More

CC2D1A Antibody (SR1124), Novus Biologicals™

Product Code. 18653215 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
Product Code. Quantity unitSize
18653215 100 μg 100µL
18654475 50 μg 50µL
2 options
This item is not returnable. View return policy

Product Code. 18653215

Brand: Novus Biologicals NBP322397100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CC2D1A Monoclonal antibody specifically detects CC2D1A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CC2D1A
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone SR1124
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Akt kinase-interacting protein 1, coiled-coil and C2 domain containing 1A, coiled-coil and C2 domain-containing protein 1A, Five repressor element under dual repression-binding protein 1, FLJ20241, FLJ41160, FRE under dual repression-binding protein 1, Freud-1, mental retardation, nonsyndromic, autosomal recessive, 3, MRT3Freud-1/Aki1, Putative NF-kappa-B-activating protein 023N, putative NFkB activating protein
Host Species Rabbit
Immunogen A synthesized peptide derived from human CC2D1A (Uniprot #: Q6P1N0)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54862
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.